MAT047024762 Matrix Scientific

Tetra-n-butylammonium dihydrogen trifluoride

  • MDL number MFCD00145365
  • Cas Number : 99337-56-1
  • Molecular Weight : 301.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Flash Point 13
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/006519.html

Safety Information

hcodes Flammable/Corrosive
MSDS http://www.matrixscientific.com/006519.html
Availability details for
Quantity
  • Availability
  • Add to Cart