MAT047024812 Matrix Scientific

1-Naphthhydrazide

  • MDL number MFCD00016804
  • Cas Number : 43038-45-5
  • Molecular Weight : 186.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-166
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/006585.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006585.html
Availability details for
Quantity
  • Availability
  • Add to Cart