MAT047024816 Matrix Scientific

3',4'-Dihydroxyacetophenone

  • MDL number MFCD00016439
  • Cas Number : 1197-09-7
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-119
Purity 98
Storage temp. Freezer
Country of Origin USA
Boiling point 127-133� Torr
Product Link http://www.matrixscientific.com/006589.html

Safety Information

hcodes Irritant/Store in Freezer
MSDS http://www.matrixscientific.com/006589.html
Availability details for
Quantity
  • Availability
  • Add to Cart