MAT047024834 Matrix Scientific

2-[1-Methyl-5-(trifluoromethyl)pyrazol-3-yl]-thiophene-5-sulfonyl chloride

  • MDL number MFCD00052283
  • Cas Number : 166964-31-4
  • Molecular Weight : 330.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-80
Country of Origin USA
Product Link http://www.matrixscientific.com/006617.html

Safety Information

hcodes Corrosive/Moisture Sensitive/Refrigerate
MSDS http://www.matrixscientific.com/006617.html
Availability details for
Quantity
  • Availability
  • Add to Cart