MAT047024845 Matrix Scientific

N,N-Dibutylbenzenesulfonamide

  • MDL number MFCD00151808
  • Cas Number : 5339-59-3
  • Molecular Weight : 269.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Boiling point 211-215� Torr
Product Link http://www.matrixscientific.com/006627.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006627.html
Availability details for
Quantity
  • Availability
  • Add to Cart