MAT047024849 Matrix Scientific

Iodomesitylene

  • MDL number MFCD00013707
  • Cas Number : 4028-63-1
  • Molecular Weight : 246.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 31
Purity 99
Storage temp. Ambient/dark
Country of Origin USA
Product Link http://www.matrixscientific.com/006635.html

Safety Information

hcodes Light Sensitive
MSDS http://www.matrixscientific.com/006635.html
Availability details for
Quantity
  • Availability
  • Add to Cart