MAT047024864 Matrix Scientific

4-n-Butoxyphenylacetonitrile

  • MDL number MFCD00049008
  • Cas Number : 38746-93-9
  • Molecular Weight : 189.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1
Country of Origin USA
Boiling point 148�m
Product Link http://www.matrixscientific.com/006661.html

Safety Information

hcodes Irritant/Irritant-Harmful
MSDS http://www.matrixscientific.com/006661.html
Availability details for
Quantity
  • Availability
  • Add to Cart