MAT047024882 Matrix Scientific

4-Iodobenzhydrazide

  • MDL number MFCD00270107
  • Cas Number : 39115-95-2
  • Molecular Weight : 262.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168-170
Country of Origin USA
Product Link http://www.matrixscientific.com/006684.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006684.html
Availability details for
Quantity
  • Availability
  • Add to Cart