MAT047024889 Matrix Scientific

4-Iodobenzophenone

  • MDL number MFCD00270109
  • Cas Number : 6136-66-9
  • Molecular Weight : 308.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-101
Purity 98
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/006692.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006692.html
Availability details for
Quantity
  • Availability
  • Add to Cart