MAT047024922 Matrix Scientific

Bromodifluoroacetic acid

  • MDL number MFCD02183519
  • Cas Number : 354-08-5
  • Molecular Weight : 174.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-113
Purity 97
Storage temp. Ambient
Country of Origin USA
Boiling point 155-160
Product Link http://www.matrixscientific.com/006740.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/006740.html
Availability details for
Quantity
  • Availability
  • Add to Cart