MAT047024932 Matrix Scientific

4-Aminomethylphenylboronic acid hydrochloride

  • MDL number MFCD01632199
  • Cas Number : 75705-21-4
  • Molecular Weight : 187.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 240-250
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/006751.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/006751.html
Availability details for
Quantity
  • Availability
  • Add to Cart