MAT047024939 Matrix Scientific

4-Bromobenzenedimethylsulfonamide

  • MDL number MFCD00017834
  • Cas Number : 707-60-8
  • Molecular Weight : 264.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-108
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/006760.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/006760.html
Availability details for
Quantity
  • Availability
  • Add to Cart