MAT047024949 Matrix Scientific

Copper(I) thiophene-2-carboxylate

  • MDL number MFCD02183524
  • Cas Number : 68986-76-5
  • Molecular Weight : 190.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Flash Point 260
Storage temp. Refrigerate/argon/dark
Country of Origin USA
Product Link http://www.matrixscientific.com/006771.html

Safety Information

hcodes Irritant/Light Sensitive/Air Sensitive/Refrigerate/Stored under Argon
MSDS http://www.matrixscientific.com/006771.html
Availability details for
Quantity
  • Availability
  • Add to Cart