MAT047024954 Matrix Scientific

1,1,1,2,2-Pentafluoro-6,6-dimethyl-3,5-heptanedione

  • MDL number MFCD00045109
  • Cas Number : 2145-68-8
  • Molecular Weight : 246.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 65�mm
Product Link http://www.matrixscientific.com/001569.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001569.html
Availability details for
Quantity
  • Availability
  • Add to Cart