MAT047024974 Matrix Scientific

2,6-Dichloro-4-methylsulfonylphenol

  • MDL number MFCD00216500
  • Cas Number : 20951-05-7
  • Molecular Weight : 241.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 223-224
Country of Origin USA
Product Link http://www.matrixscientific.com/006799.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006799.html
Availability details for
Quantity
  • Availability
  • Add to Cart