MAT047024993 Matrix Scientific

1-(4-Chlorophenyl)-1,2-propandione

  • MDL number MFCD01941300
  • Cas Number : 10557-21-8
  • Molecular Weight : 182.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 22-23
Density 1.2700/20
Country of Origin USA
Boiling point 54-59�03 Torr
Product Link http://www.matrixscientific.com/006816.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006816.html
Availability details for
Quantity
  • Availability
  • Add to Cart