MAT047024999 Matrix Scientific

2-Methylsulfonyl-1-(3-trifluoromethylphenyl)-ethanone

  • MDL number MFCD00665597
  • Cas Number : 128306-96-7
  • Molecular Weight : 266.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-94
Country of Origin USA
Product Link http://www.matrixscientific.com/006821.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006821.html
Availability details for
Quantity
  • Availability
  • Add to Cart