MAT047025002 Matrix Scientific

Methyl 6-(trifluoromethyl)nicotinoylacetate

  • MDL number MFCD01862654
  • Cas Number : 386704-15-0
  • Molecular Weight : 247.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-62
Country of Origin USA
Product Link http://www.matrixscientific.com/006824.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006824.html
Availability details for
Quantity
  • Availability
  • Add to Cart