MAT047025004 Matrix Scientific

6-(Trifluoromethyl)pyridine-3-amidinehydrochloride

  • MDL number MFCD01862652
  • Cas Number : 221313-11-7
  • Molecular Weight : 225.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191-193
Country of Origin USA
Product Link http://www.matrixscientific.com/006826.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006826.html
Availability details for
Quantity
  • Availability
  • Add to Cart