MAT047025022 Matrix Scientific

4-Methylsulfonylbenzonitrile

  • MDL number MFCD00216489
  • Cas Number : 22821-76-7
  • Molecular Weight : 181.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-142
Country of Origin USA
Product Link http://www.matrixscientific.com/006842.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006842.html
Availability details for
Quantity
  • Availability
  • Add to Cart