MAT047025026 Matrix Scientific

2-(5-Methoxy-2-nitrophenyl)malondialdehydemonohydrate

  • MDL number MFCD00216510
  • Cas Number : 205692-61-1
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-108�c)
Country of Origin USA
Product Link http://www.matrixscientific.com/006846.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006846.html
Availability details for
Quantity
  • Availability
  • Add to Cart