MAT047025046 Matrix Scientific

3-Formyl-2-methyl-5-nitroindole

  • MDL number MFCD00216481
  • Cas Number : 3558-17-6
  • Molecular Weight : 204.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 305-307�c)
Country of Origin USA
Product Link http://www.matrixscientific.com/006864.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006864.html
Availability details for
Quantity
  • Availability
  • Add to Cart