MAT047025077 Matrix Scientific

Coumarin-6-carboxaldehyde

  • MDL number MFCD01941322
  • Cas Number : 51690-26-7
  • Molecular Weight : 174.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-194
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/006891.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006891.html
Availability details for
Quantity
  • Availability
  • Add to Cart