MAT047025083 Matrix Scientific

1-(3,5-Dichlorophenyl)-1,2-propandione

  • MDL number MFCD01941316
  • Cas Number : 386715-49-7
  • Molecular Weight : 217.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-46
Country of Origin USA
Product Link http://www.matrixscientific.com/006897.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006897.html
Availability details for
Quantity
  • Availability
  • Add to Cart