MAT047025085 Matrix Scientific

1-(5-Bromopyridin-3-yl)-2-methylsulfonyl-ethanone

  • MDL number MFCD01941313
  • Cas Number : 386715-50-0
  • Molecular Weight : 278.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-161
Country of Origin USA
Product Link http://www.matrixscientific.com/006899.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006899.html
Availability details for
Quantity
  • Availability
  • Add to Cart