MAT047025090 Matrix Scientific

2-Methylsulfonyl-1-(4-trifluoromethylphenyl)-ethanone

  • MDL number MFCD01941309
  • Cas Number : 386715-52-2
  • Molecular Weight : 266.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-102
Country of Origin USA
Product Link http://www.matrixscientific.com/006903.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006903.html
Availability details for
Quantity
  • Availability
  • Add to Cart