MAT047025105 Matrix Scientific

3-[2,3-Dihydro-1-(1H)-indole]propanoic acid

  • MDL number MFCD06247004
  • Cas Number : 859190-83-3
  • Molecular Weight : 191.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-77
Country of Origin USA
Product Link http://www.matrixscientific.com/006917.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006917.html
Availability details for
Quantity
  • Availability
  • Add to Cart