MAT047025123 Matrix Scientific

2-Methyl-1,8-naphthyridine-3-carboxylic acid

  • MDL number MFCD00202882
  • Cas Number : 387350-60-9
  • Molecular Weight : 188.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185
Country of Origin USA
Product Link http://www.matrixscientific.com/006933.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006933.html
Availability details for
Quantity
  • Availability
  • Add to Cart