MAT047025128 Matrix Scientific

2-(Trifluoromethyl)-1,6-naphthyridine-3-carboxylic acid

  • MDL number MFCD00202962
  • Cas Number : 240408-97-3
  • Molecular Weight : 242.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 233-235
Country of Origin USA
Product Link http://www.matrixscientific.com/006938.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006938.html
Availability details for
Quantity
  • Availability
  • Add to Cart