MAT047025133 Matrix Scientific

2-(3-Hydroxycarbonyl-2-nitrophenyl)-malondialdehyde

  • MDL number MFCD00216502
  • Cas Number : 205680-83-7
  • Molecular Weight : 237.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215-217�c)
Country of Origin USA
Product Link http://www.matrixscientific.com/006943.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006943.html
Availability details for
Quantity
  • Availability
  • Add to Cart