MAT047025134 Matrix Scientific

2-(5-Hydroxycarbonyl-2-nitrophenyl)-malondialdehyde monohydrate

  • MDL number MFCD00216507
  • Cas Number : 205680-84-8
  • Molecular Weight : 255.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214-216�c)
Country of Origin USA
Product Link http://www.matrixscientific.com/006944.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006944.html
Availability details for
Quantity
  • Availability
  • Add to Cart