MAT047025137 Matrix Scientific

Methyl 5-bromonicotinoylacetate

  • MDL number MFCD00216527
  • Cas Number : 205985-97-3
  • Molecular Weight : 258.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-94
Country of Origin USA
Product Link http://www.matrixscientific.com/006947.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006947.html
Availability details for
Quantity
  • Availability
  • Add to Cart