MAT047025154 Matrix Scientific

2,3-Dihydro-1-(methylsulfonyl)-(1H)-indole-5-amine

  • MDL number MFCD01319362
  • Cas Number : 299921-01-0
  • Molecular Weight : 212.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-117
Country of Origin USA
Product Link http://www.matrixscientific.com/006963.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006963.html
Availability details for
Quantity
  • Availability
  • Add to Cart