MAT047025156 Matrix Scientific

alpha-Methyl-4-(methylsulfonyl)benzylamine

  • MDL number MFCD01319250
  • Cas Number : 387350-90-5
  • Molecular Weight : 199.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-65
Country of Origin USA
Product Link http://www.matrixscientific.com/006965.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006965.html
Availability details for
Quantity
  • Availability
  • Add to Cart