MAT047025176 Matrix Scientific

1-Acetyl-4-cyclopropylaminopiperidine

  • MDL number MFCD01941331
  • Cas Number : 387358-46-5
  • Molecular Weight : 182.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 130�15mm
Product Link http://www.matrixscientific.com/006982.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006982.html
Availability details for
Quantity
  • Availability
  • Add to Cart