MAT047025179 Matrix Scientific

5-Chloro-2,3-dihydro-(1H)-indole

  • MDL number MFCD01941330
  • Cas Number : 25658-80-4
  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Refrigerate
Country of Origin USA
Boiling point 93-98�4 Torr
Product Link http://www.matrixscientific.com/006985.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/006985.html
Availability details for
Quantity
  • Availability
  • Add to Cart