MAT047025181 Matrix Scientific

4-Cyclopropylamino-1-methylpiperidine

  • MDL number MFCD01941329
  • Cas Number : 387358-48-7
  • Molecular Weight : 154.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 36�05mm
Product Link http://www.matrixscientific.com/006987.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006987.html
Availability details for
Quantity
  • Availability
  • Add to Cart