MAT047025244 Matrix Scientific

5-(Diisobutylamino)amylamine

  • MDL number MFCD00048343
  • Cas Number : 209803-39-4
  • Molecular Weight : 214.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 95�mm
Product Link http://www.matrixscientific.com/007225.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/007225.html
Availability details for
Quantity
  • Availability
  • Add to Cart