MAT047025270 Matrix Scientific

2-(Diallylamino)ethylamine

  • MDL number MFCD03452794
  • Cas Number : 25077-82-1
  • Molecular Weight : 140.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 0.8523/20
Country of Origin USA
Boiling point 79� Torr
Product Link http://www.matrixscientific.com/007545.html

Safety Information

hcodes Corrosive/Irritant
MSDS http://www.matrixscientific.com/007545.html
Availability details for
Quantity
  • Availability
  • Add to Cart