MAT047025285 Matrix Scientific

5-[4-(Trifluoromethyl)phenyl]isoxazole

  • MDL number MFCD02183551
  • Cas Number : 387824-55-7
  • Molecular Weight : 213.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007041.html

Safety Information

MSDS http://www.matrixscientific.com/007041.html
Availability details for
Quantity
  • Availability
  • Add to Cart