MAT047025287 Matrix Scientific

4-Benzenesulfonylthiophene-2-sulfonyl chloride

  • MDL number MFCD00052138
  • Cas Number : 160233-28-3
  • Molecular Weight : 322.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-103
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/007043.html

Safety Information

hcodes Corrosive/Moisture Sensitive
MSDS http://www.matrixscientific.com/007043.html
Availability details for
Quantity
  • Availability
  • Add to Cart