MAT047025297 Matrix Scientific

2-Amino-4,5-dimethoxybenzonitrile

  • MDL number MFCD00058682
  • Cas Number : 26961-27-3
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-94
Purity 99
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/007054.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/007054.html
Availability details for
Quantity
  • Availability
  • Add to Cart