MAT047025311 Matrix Scientific

2-Chloro-5-nitrocinnamic acid

  • MDL number MFCD00063312
  • Cas Number : 36015-19-7
  • Molecular Weight : 227.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220-221�ec)
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/007070.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007070.html
Availability details for
Quantity
  • Availability
  • Add to Cart