MAT047025379 Matrix Scientific

4-Bromo-1-chloro-2-fluorobenzene

  • MDL number MFCD00042519
  • Cas Number : 60811-18-9
  • Molecular Weight : 209.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.71
Country of Origin USA
Boiling point 62-64/9mm
Product Link http://www.matrixscientific.com/001609.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001609.html
Availability details for
Quantity
  • Availability
  • Add to Cart