MAT047025437 Matrix Scientific

6-Chloro-7-hydroxy-4-(trifluoromethyl)coumarin

  • MDL number MFCD00068095
  • Cas Number : 119179-66-7
  • Molecular Weight : 264.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-183
Country of Origin USA
Product Link http://www.matrixscientific.com/007211.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007211.html
Availability details for
Quantity
  • Availability
  • Add to Cart