MAT047025454 Matrix Scientific

5-(Dimethylamino)amylamine

  • MDL number MFCD00047978
  • Cas Number : 3209-46-9
  • Molecular Weight : 130.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 50� Torr
Product Link http://www.matrixscientific.com/007229.html

Safety Information

hcodes Corrosive/Stored under Argon
MSDS http://www.matrixscientific.com/007229.html
Availability details for
Quantity
  • Availability
  • Add to Cart