MAT047025456 Matrix Scientific

N-Ethyl-N-butylethylenediamine

  • MDL number MFCD00047953
  • Cas Number : 19435-65-5
  • Molecular Weight : 144.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 0.8197/20
Country of Origin USA
Boiling point 165-167
Product Link http://www.matrixscientific.com/007231.html

Safety Information

hcodes Irritant-Harmful
MSDS http://www.matrixscientific.com/007231.html
Availability details for
Quantity
  • Availability
  • Add to Cart