MAT047025458 Matrix Scientific

2-Aminoimidazole sulfate

  • MDL number MFCD00050811
  • Cas Number : 1450-93-7
  • Molecular Weight : 181.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 252�c)
Purity 98
Storage temp. Freezer
Country of Origin USA
Product Link http://www.matrixscientific.com/007232.html

Safety Information

hcodes Irritant/Store in Freezer
MSDS http://www.matrixscientific.com/007232.html
Availability details for
Quantity
  • Availability
  • Add to Cart