MAT047025462 Matrix Scientific

4-Bromo-2,2-diphenylbutyric acid

  • MDL number MFCD01321300
  • Cas Number : 37742-98-6
  • Molecular Weight : 319.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-132
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/007236.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007236.html
Availability details for
Quantity
  • Availability
  • Add to Cart