MAT047025468 Matrix Scientific

2-Bromo-3-chloro-5-(trifluoromethyl)pyridine

  • MDL number MFCD00153072
  • Cas Number : 75806-84-7
  • Molecular Weight : 260.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 80�mm
Product Link http://www.matrixscientific.com/001622.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001622.html
Availability details for
Quantity
  • Availability
  • Add to Cart